Assay Details
Target Gene Details
Entrez Gene ID: | 22941 |
Gene Name: | SH3 and multiple ankyrin repeat domains 2 |
Gene Aliases: |
AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3 |
Location: |
Chr.11:70467856-71252724 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHANK2 | NM_012309.4 | NP_036441.2 | ||
| NM_133266.4 | NP_573573.2 | |||
| NR_110766.1 | ||||
| XM_005277932.3 | XP_005277989.1 | |||
| XM_017017387.1 | XP_016872876.1 | |||
| XM_017017388.1 | XP_016872877.1 | |||
| XM_017017389.1 | XP_016872878.1 | |||
| XM_017017390.1 | XP_016872879.1 | |||
| AB208025.1 | ||||
| AB208026.1 | ||||
| AB208027.1 | ||||
| AF141901.1 | AAF02496.1 | |||
| BC093885.1 | AAH93885.1 | |||
| BC112097.1 | AAI12098.1 | |||
| BC114484.1 | AAI14485.1 | |||
| BC143389.1 | ||||
| DQ152234.1 | AAZ77790.1 |
Target Gene Details
Entrez Gene ID: | 100874198 |
Gene Name: | SHANK2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:70631094-70635490 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHANK2-AS1 | NR_046567.1 | |||
| AK097446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818850 | Chr.11:70630705 - 70650536 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map