Assay Details
Target Gene Details
Entrez Gene ID: | 960 |
Gene Name: | CD44 molecule (Indian blood group) |
Gene Aliases: |
CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1 |
Location: |
Chr.11:35138870-35232402 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759818 | Chr.11:35008386 - 35217031 on Build GRCh38 | Loss |
![]() ![]() |
nsv1042014 | Chr.11:34919190 - 37195907 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3625922 | Chr.11:35148319 - 35190916 on Build GRCh38 | Loss |
![]() |
nsv951602 | Chr.11:35160554 - 35175653 on Build GRCh38 | Duplication |
![]() |
nsv508623 | Chr.11:35073386 - 35181816 on Build GRCh38 | Deletion |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
