Assay Details
Target Gene Details
Entrez Gene ID: | 57689 |
Gene Name: | leucine rich repeat containing 4C |
Gene Aliases: |
NGL-1, NGL1 |
Location: |
Chr.11:40107066-41460212 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC4C | XM_011520238.2 | 8 | 5608 | XP_011518540.1 |
| XM_011520239.2 | 9 | 5674 | XP_011518541.1 | |
| XM_011520240.2 | 7 | 5528 | XP_011518542.1 | |
| XM_011520241.2 | 4 | 6243 | XP_011518543.1 | |
| XM_011520242.2 | 4 | 6229 | XP_011518544.1 | |
| XM_011520243.2 | 7 | 5555 | XP_011518545.1 | |
| XM_011520244.2 | 6 | 5475 | XP_011518546.1 | |
| XM_017018070.1 | 8 | 5604 | XP_016873559.1 | |
| XM_017018071.1 | 8 | 5594 | XP_016873560.1 | |
| XM_017018072.1 | 8 | 5457 | XP_016873561.1 | |
| XM_017018073.1 | 6 | 5316 | XP_016873562.1 | |
| XM_017018074.1 | 4 | 6224 | XP_016873563.1 | |
| XM_017018075.1 | 5 | 5263 | XP_016873564.1 | |
| XM_017018076.1 | 3 | 6190 | XP_016873565.1 | |
| XM_017018077.1 | 6 | 5343 | XP_016873566.1 | |
| XM_017018078.1 | 5 | 6309 | XP_016873567.1 | |
| XM_017018079.1 | 6 | 5471 | XP_016873568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv65n21 | Chr.11:40049473 - 40249892 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72885365] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map