Assay Details
Target Gene Details
Entrez Gene ID: | 79899 |
Gene Name: | proline rich 5 like |
Gene Aliases: |
PROTOR2 |
Location: |
Chr.11:36296175-36465204 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR5L | NM_001160167.1 | NP_001153639.1 | ||
| AK055042.1 | ||||
| DA269290.1 | ||||
| DA344861.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042014 | Chr.11:34919190 - 37195907 on Build GRCh38 | Loss |
|
| nsv467791 | Chr.11:36086947 - 36977835 on Build GRCh38 | Gain |
|
| esv3309539 | Chr.11:36318476 - 36318477 on Build GRCh38 | Mobile Element Insertion |
|
| esv2422305 | Chr.11:35928148 - 36967365 on Build GRCh38 | Duplication |
|
| esv3304080 | Chr.11:36318488 - 36318489 on Build GRCh38 | Mobile Element Insertion |
|
| esv3425565 | Chr.11:36318484 - 36318502 on Build GRCh38 | Insertion |
|
| dgv341e212 | Chr.11:36314958 - 36336812 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map