Assay Details
Target Gene Details
Entrez Gene ID: | 10714 |
Gene Name: | DNA polymerase delta 3, accessory subunit |
Gene Aliases: |
P66, P68, PPP1R128 |
Location: |
Chr.11:74592530-74669341 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLD3 | NM_006591.2 | NP_006582.1 | ||
| NR_046409.1 | ||||
| NR_046410.1 | ||||
| XM_005273716.1 | XP_005273773.1 | |||
| XM_011544734.2 | XP_011543036.1 | |||
| AK302677.1 | ||||
| AK316301.1 | ||||
| AK316370.1 | ||||
| BC032636.1 | ||||
| BC092454.1 | AAH92454.1 | |||
| BC108908.1 | AAI08909.1 | |||
| BC108909.1 | AAI08910.1 | |||
| D26018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34979 | Chr.11:74167643 - 74822407 on Build GRCh38 | Gain |
|
| esv2759837 | Chr.11:74131564 - 74846552 on Build GRCh38 | Gain |
|
| dgv209e214 | Chr.11:74225600 - 74828943 on Build GRCh38 | Gain |
|
| nsv825994 | Chr.11:74225622 - 74823410 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map