Assay Details
Target Gene Details
Entrez Gene ID: | 51075 |
Gene Name: | thioredoxin related transmembrane protein 2 |
Gene Aliases: |
CGI-31, PDIA12, PIG26, TXNDC14 |
Location: |
Chr.11:57712523-57740973 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMX2 | NM_001144012.2 | NP_001137484.1 | ||
| NM_015959.3 | NP_057043.1 | |||
| NR_026593.2 | ||||
| NR_037645.1 | ||||
| AF059753.1 | AAG43122.1 | |||
| AF132965.1 | AAD27740.1 | |||
| AK075359.1 | BAC11569.1 | |||
| AK223058.1 | BAD96778.1 | |||
| AK297688.1 | ||||
| AK298065.1 | ||||
| AK312990.1 | ||||
| AL037895.1 | ||||
| AY358501.1 | AAQ88865.1 | |||
| AY451236.1 | ||||
| AY451237.1 | AAS66642.1 | |||
| BC000666.1 | AAH00666.1 | |||
| BC093062.1 | AAH93062.1 | |||
| BQ686696.1 | ||||
| CD690929.1 |
Target Gene Details
Entrez Gene ID: | 100528016 |
Gene Name: | TMX2-CTNND1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:57712523-57819180 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMX2-CTNND1 | NR_037646.1 | |||
| AB002382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3579728 | Chr.11:57686071 - 57726599 on Build GRCh38 | Loss |
|
| nsv832163 | Chr.11:57531881 - 57713578 on Build GRCh38 | Gain |
|
| nsv508633 | Chr.11:57652112 - 57792573 on Build GRCh38 | Deletion |
|
| nsv975188 | Chr.11:57712848 - 57726726 on Build GRCh38 | Duplication |
|
| nsv832164 | Chr.11:57640144 - 57811353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map