Assay Details
Target Gene Details
Entrez Gene ID: | 22877 |
Gene Name: | MLX interacting protein |
Gene Aliases: |
MIR, MONDOA, bHLHe36 |
Location: |
Chr.12:122078728-122147347 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLXIP | NM_014938.5 | 1 | 124 | NP_055753.3 |
| XM_006719290.3 | 1 | 108 | XP_006719353.1 | |
| XM_006719291.3 | 1 | 108 | XP_006719354.1 | |
| XM_006719292.3 | 1 | 108 | XP_006719355.1 | |
| XM_006719293.3 | 1 | 108 | XP_006719356.1 | |
| AF312918.1 | AAG34121.1 | |||
| BC151841.1 | 1 | 106 | ||
| BM461435.1 | 1 | 160 |
More Information
Additional Information:
For this assay, SNP(s) [rs183909049] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map