Assay Details
Target Gene Details
Entrez Gene ID: | 84749 |
Gene Name: | ubiquitin specific peptidase 30 |
Gene Aliases: |
- |
Location: |
Chr.12:109023089-109088026 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP30 | NM_001301175.1 | NP_001288104.1 | ||
| AK097830.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422361 | Chr.12:108792678 - 109118360 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117993355] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map