Assay Details
Target Gene Details
Entrez Gene ID: | 10867 |
Gene Name: | tetraspanin 9 |
Gene Aliases: |
NET-5, NET5, PP1057 |
Location: |
Chr.12:3077355-3286564 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN9 | NM_001168320.1 | NP_001161792.1 | ||
| NM_006675.4 | NP_006666.1 | |||
| XM_011520912.2 | XP_011519214.1 | |||
| AF089749.1 | AAC35859.1 | |||
| AK027366.1 | ||||
| AK223179.1 | BAD96899.1 | |||
| BC071881.1 | AAH71881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832312 | Chr.12:3133005 - 3358034 on Build GRCh38 | Loss |
|
| nsv468976 | Chr.12:3136424 - 3146196 on Build GRCh38 | Loss |
|
| nsv527090 | Chr.12:3131129 - 3146196 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map