Assay Details
Target Gene Details
Entrez Gene ID: | 387890 |
Gene Name: | transmembrane protein 233 |
Gene Aliases: |
DSPB2, IFITMD2 |
Location: |
Chr.12:119592123-119654455 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM233 | NM_001136534.1 | NP_001130006.1 | ||
| XM_005253880.3 | XP_005253937.1 | |||
| XM_005253881.3 | XP_005253938.1 | |||
| XM_011538328.2 | XP_011536630.1 | |||
| XM_011538329.2 | XP_011536631.1 | |||
| XM_011538330.2 | XP_011536632.1 | |||
| XM_011538331.2 | XP_011536633.1 | |||
| AK296289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630909 | Chr.12:119587649 - 119641066 on Build GRCh38 | Gain |
|
| nsv832529 | Chr.12:119425573 - 119633841 on Build GRCh38 | Loss |
|
| esv3630908 | Chr.12:119581656 - 119611233 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map