Assay Details
Target Gene Details
Entrez Gene ID: | 23457 |
Gene Name: | ATP binding cassette subfamily B member 9 |
Gene Aliases: |
EST122234, TAPL |
Location: |
Chr.12:122918708-122975160 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss |
|
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |
|

Genomic Map