Assay Details
Target Gene Details
Entrez Gene ID: | 1431 |
Gene Name: | citrate synthase |
Gene Aliases: |
- |
Location: |
Chr.12:56271699-56300391 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CS | NM_004077.2 | NP_004068.2 | ||
| AF047042.1 | AAC25560.1 | |||
| AF053631.1 | AAQ13428.1 | |||
| AF147410.1 | ||||
| AK074956.1 | BAC11314.1 | |||
| AK095856.1 | ||||
| AK293263.1 | ||||
| AK296249.1 | ||||
| AK307625.1 | ||||
| BC000105.1 | AAH00105.3 | |||
| BC010106.1 | AAH10106.1 | |||
| BC072016.1 | AAH72016.1 | |||
| BX640838.1 | CAE45911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map