Assay Details
Target Gene Details
Entrez Gene ID: | 2967 |
Gene Name: | general transcription factor IIH subunit 3 |
Gene Aliases: |
BTF2, P34, TFB4, TFIIH |
Location: |
Chr.12:123633739-123662604 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2H3 | NM_001271866.1 | NP_001258795.1 | ||
| NM_001271867.1 | NP_001258796.1 | |||
| NM_001271868.1 | NP_001258797.1 | |||
| NM_001516.4 | NP_001507.2 | |||
| XM_017019228.1 | XP_016874717.1 | |||
| AK298128.1 | ||||
| AK313200.1 | ||||
| AL558542.3 | ||||
| BC031030.1 | AAH31030.1 | |||
| BC039726.2 | AAH39726.1 | |||
| BC047868.2 | AAH47868.2 | |||
| BC065250.1 | AAH65250.1 | |||
| BG720598.1 | ||||
| DA953586.1 | ||||
| DB170244.1 | ||||
| Z30093.1 | CAA82909.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1566n100 | Chr.12:123610211 - 123711309 on Build GRCh38 | Gain |
|
| esv2759918 | Chr.12:123625575 - 123711309 on Build GRCh38 | Gain |
|
| nsv455733 | Chr.12:123635436 - 123711309 on Build GRCh38 | Gain |
|
| nsv442292 | Chr.12:123630500 - 123711307 on Build GRCh38 | Gain |
|
| esv35148 | Chr.12:123634573 - 123711573 on Build GRCh38 | Gain |
|
| dgv1567n100 | Chr.12:123634608 - 123694870 on Build GRCh38 | Gain |
|
| dgv309e214 | Chr.12:123628443 - 123712471 on Build GRCh38 | Gain |
|
| dgv1568n100 | Chr.12:123637829 - 123678203 on Build GRCh38 | Loss |
|
| nsv1042181 | Chr.12:123552853 - 123668403 on Build GRCh38 | Gain |
|
| nsv1051385 | Chr.12:123642456 - 123675474 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map