Assay Details
Target Gene Details
Entrez Gene ID: | 144423 |
Gene Name: | glycosyltransferase 1 domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.12:128853323-128984964 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLT1D1 | NM_144669.2 | NP_653270.1 | ||
| NR_133646.1 | ||||
| XM_005253553.4 | XP_005253610.2 | |||
| XM_011537956.2 | XP_011536258.2 | |||
| XM_011537957.2 | XP_011536259.1 | |||
| XM_017018855.1 | XP_016874344.1 | |||
| XM_017018856.1 | XP_016874345.1 | |||
| XM_017018857.1 | XP_016874346.1 | |||
| XM_017018858.1 | XP_016874347.1 | |||
| XM_017018859.1 | XP_016874348.1 | |||
| XM_017018860.1 | XP_016874349.1 | |||
| XM_017018861.1 | XP_016874350.1 | |||
| AK056540.1 | BAB71209.1 | |||
| AK295333.1 | ||||
| BC043528.1 | AAH43528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051433 | Chr.12:128679360 - 128888555 on Build GRCh38 | Gain |
|
| esv3631153 | Chr.12:128855742 - 128901072 on Build GRCh38 | Gain |
|
| esv2759920 | Chr.12:128587760 - 128896765 on Build GRCh38 | Loss |
|
| nsv1040831 | Chr.12:128749443 - 129516029 on Build GRCh38 | Gain |
|
| esv3581993 | Chr.12:128679608 - 128885566 on Build GRCh38 | Gain |
|
| nsv832549 | Chr.12:128739165 - 128936554 on Build GRCh38 | Loss |
|
| nsv560689 | Chr.12:128840393 - 129063666 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map