Assay Details
Target Gene Details
Entrez Gene ID: | 160492 |
Gene Name: | lamin tail domain containing 1 |
Gene Aliases: |
IFLTD1, PAS1C1 |
Location: |
Chr.12:25476082-25648604 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMNTD1 | NM_001145728.2 | NP_001139200.1 | ||
| NM_001256266.1 | NP_001243195.1 | |||
| NM_152590.3 | NP_689803.2 | |||
| XM_011520575.1 | XP_011518877.1 | |||
| AK093323.1 | BAC04132.1 | |||
| AK302292.1 | ||||
| BC037957.1 | AAH37957.1 | |||
| DW009862.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2665786 | Chr.12:25494238 - 25501989 on Build GRCh38 | Deletion |
|
| esv3548905 | Chr.12:25494067 - 25502566 on Build GRCh38 | Deletion |
|
| esv3628898 | Chr.12:25494286 - 25501874 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map