Assay Details
Target Gene Details
Entrez Gene ID: | 253724 |
Gene Name: | tetratricopeptide repeat domain 41, pseudogene |
Gene Aliases: |
GNN, GNNP |
Location: |
Chr.12:103843749-103930211 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC41P | NR_027249.1 | 16 | 3948 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560039 | Chr.12:103783890 - 104061562 on Build GRCh38 | Gain |
|
| esv34031 | Chr.12:103720499 - 103904768 on Build GRCh38 | Loss |
|

Genomic Map