Assay Details
Target Gene Details
Entrez Gene ID: | 10301 |
Gene Name: | deleted in lymphocytic leukemia 1 (non-protein coding) |
Gene Aliases: |
BCMS, BCMS1, DLB1, DLEU2, LEU1, LEU2, LINC00021, NCRNA00021, XTP6 |
Location: |
Chr.13:50082169-50528643 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLEU1 | NR_109974.1 | |||
| AJ412026.1 | ||||
| AJ412027.1 | ||||
| AJ412028.1 | ||||
| AJ412029.1 | ||||
| AJ412031.1 | ||||
| AJ412032.1 | ||||
| AJ412035.1 | ||||
| AJ412036.1 | ||||
| AJ412037.1 | ||||
| AJ412043.1 | ||||
| AJ412055.1 | ||||
| AJ412056.1 | ||||
| AJ412057.1 | ||||
| AJ412058.1 | ||||
| AJ412059.1 | ||||
| AJ412060.1 | ||||
| AJ412061.1 | ||||
| AJ412062.1 | ||||
| AJ412063.1 |
Target Gene Details
Entrez Gene ID: | 103689915 |
Gene Name: | DLEU1 antisense RNA 1 |
Gene Aliases: |
LINC01308 |
Location: |
Chr.13:50520933-50527449 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLEU1-AS1 | NR_125753.1 | |||
| BM264234.1 | ||||
| DA176728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580637 | Chr.13:50006792 - 50829882 on Build GRCh38 | Loss |
|
| nsv1045237 | Chr.13:50402424 - 50528250 on Build GRCh38 | Loss |
|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
| nsv1035151 | Chr.13:49939399 - 50593714 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73184428] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map