Assay Details
Target Gene Details
Entrez Gene ID: | 51084 |
Gene Name: | crystallin lambda 1 |
Gene Aliases: |
GDH, HEL30, lambda-CRY |
Location: |
Chr.13:20403667-20525873 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYL1 | NM_015974.2 | NP_057058.2 | ||
| XM_005266415.3 | XP_005266472.1 | |||
| XM_017020624.1 | XP_016876113.1 | |||
| AF077049.1 | AAD27782.1 | |||
| AF087898.1 | AAP97197.1 | |||
| AF160216.1 | AAF67017.1 | |||
| AK024041.1 | ||||
| BC008562.2 | ||||
| BC071810.1 | ||||
| BC119660.1 | ||||
| BC119661.1 | ||||
| BQ006686.1 | ||||
| EU794604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34984 | Chr.13:20398561 - 20542402 on Build GRCh38 | Loss |
|
| dgv1612n100 | Chr.13:20222652 - 20530273 on Build GRCh38 | Loss |
|
| esv3892301 | Chr.13:20401130 - 20514578 on Build GRCh38 | Loss |
|
| esv3631431 | Chr.13:20410739 - 20423024 on Build GRCh38 | Loss |
|
| dgv221n27 | Chr.13:20398414 - 20521581 on Build GRCh38 | Loss |
|
| dgv3072n54 | Chr.13:20216603 - 20521581 on Build GRCh38 | Loss |
|
| nsv1046946 | Chr.13:19730301 - 20507454 on Build GRCh38 | Gain |
|
| esv2759925 | Chr.13:20398583 - 20650339 on Build GRCh38 | Loss |
|
| esv2751132 | Chr.13:20217943 - 20546665 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map