Assay Details
Target Gene Details
Entrez Gene ID: | 10301 |
Gene Name: | deleted in lymphocytic leukemia 1 (non-protein coding) |
Gene Aliases: |
BCMS, BCMS1, DLB1, DLEU2, LEU1, LEU2, LINC00021, NCRNA00021, XTP6 |
Location: |
Chr.13:50082169-50528643 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLEU1 | NR_002605.2 | |||
| NR_109973.1 | ||||
| NR_109974.1 | ||||
| AF490255.1 | ||||
| AJ412023.1 | ||||
| AJ412024.1 | ||||
| AJ412026.1 | ||||
| AJ412038.1 | ||||
| AJ412039.1 | ||||
| AJ412054.1 | ||||
| AJ412055.1 | ||||
| AJ412056.1 | ||||
| AJ412057.1 | ||||
| AJ412058.1 | ||||
| AJ412059.1 | ||||
| AJ412060.1 | ||||
| AJ412061.1 | ||||
| AJ412062.1 | ||||
| AK095311.1 | ||||
| AK312167.1 | ||||
| BC020692.1 | ||||
| CR450325.1 | ||||
| DB444728.1 |
Target Gene Details
Entrez Gene ID: | 8847 |
Gene Name: | deleted in lymphocytic leukemia 2 (non-protein coding) |
Gene Aliases: |
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS |
Location: |
Chr.13:49982552-50125541 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLEU2 | NR_002612.1 | |||
| AF380424.1 | ||||
| AY455757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580637 | Chr.13:50006792 - 50829882 on Build GRCh38 | Loss |
|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
| nsv951082 | Chr.13:50072065 - 50085464 on Build GRCh38 | Duplication |
|
| nsv1035151 | Chr.13:49939399 - 50593714 on Build GRCh38 | Loss |
|

Genomic Map