Assay Details
Target Gene Details
Entrez Gene ID: | 144811 |
Gene Name: | laccase domain containing 1 |
Gene Aliases: |
C13orf31 |
Location: |
Chr.13:43878834-43893932 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LACC1 | NM_001128303.1 | NP_001121775.1 | ||
| NM_153218.2 | NP_694950.2 | |||
| XM_005266261.3 | XP_005266318.1 | |||
| XM_006719766.3 | XP_006719829.1 | |||
| XM_006719767.3 | XP_006719830.1 | |||
| XM_011534933.2 | XP_011533235.1 | |||
| XM_011534934.2 | XP_011533236.1 | |||
| XM_011534935.2 | XP_011533237.1 | |||
| XM_017020394.1 | XP_016875883.1 | |||
| AK096044.1 | BAC04686.1 | |||
| BC035749.1 | AAH35749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832595 | Chr.13:43716837 - 43925163 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146148689] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map