Assay Details
Target Gene Details
Entrez Gene ID: | 348013 |
Gene Name: | transmembrane protein 255B |
Gene Aliases: |
FAM70B |
Location: |
Chr.13:113759217-113813797 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM255B | NM_182614.2 | NP_872420.1 | ||
| XM_005272413.3 | XP_005272470.1 | |||
| XM_005272415.2 | XP_005272472.1 | |||
| XM_006719986.2 | XP_006720049.1 | |||
| XM_017020558.1 | XP_016876047.1 | |||
| XM_017020559.1 | XP_016876048.1 | |||
| XM_017020560.1 | XP_016876049.1 | |||
| BC018995.2 | AAH18995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951932 | Chr.13:113782828 - 113807827 on Build GRCh38 | Deletion |
|
| nsv1043348 | Chr.13:113756321 - 113867896 on Build GRCh38 | Gain |
|
| nsv563388 | Chr.13:113754200 - 113872798 on Build GRCh38 | Loss |
|
| nsv511528 | Chr.13:113799443 - 113804564 on Build GRCh38 | Loss |
|
| nsv832727 | Chr.13:113753115 - 113918257 on Build GRCh38 | Loss |
|
| esv3582860 | Chr.13:113756381 - 113867871 on Build GRCh38 | Gain |
|
| nsv826836 | Chr.13:113753163 - 113875076 on Build GRCh38 | Loss |
|
| esv3633615 | Chr.13:113724844 - 113843481 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78826356] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map