Assay Details
Target Gene Details
Entrez Gene ID: | 79587 |
Gene Name: | cysteinyl-tRNA synthetase 2, mitochondrial (putative) |
Gene Aliases: |
COXPD27, cysRS |
Location: |
Chr.13:110641410-110713527 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARS2 | XM_006719953.2 | XP_006720016.1 | ||
| XM_017020741.1 | XP_016876230.1 | |||
| AK301907.1 |
Target Gene Details
Entrez Gene ID: | 105376684 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.13:110710644-110714090 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105376684 | XM_011521137.2 | 1 | 2229 | XP_011519439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044234 | Chr.13:110490706 - 110718233 on Build GRCh38 | Gain |
|
| nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map