Assay Details
Target Gene Details
Entrez Gene ID: | 144811 |
Gene Name: | laccase domain containing 1 |
Gene Aliases: |
C13orf31 |
Location: |
Chr.13:43878834-43893932 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LACC1 | NM_001128303.1 | 7 | 3977 | NP_001121775.1 |
| NM_153218.2 | 7 | 3813 | NP_694950.2 | |
| XM_005266261.3 | 7 | 4436 | XP_005266318.1 | |
| XM_006719766.3 | 7 | 3594 | XP_006719829.1 | |
| XM_006719767.3 | 6 | 3652 | XP_006719830.1 | |
| XM_011534933.2 | 8 | 4631 | XP_011533235.1 | |
| XM_011534934.2 | 7 | 3750 | XP_011533236.1 | |
| XM_017020394.1 | 7 | 3870 | XP_016875883.1 | |
| BX538036.1 | 3 | 2548 | CAD97980.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832595 | Chr.13:43716837 - 43925163 on Build GRCh38 | Gain |
|
| nsv524104 | Chr.13:43889273 - 43897141 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map