Assay Details
Target Gene Details
Entrez Gene ID: | 8874 |
Gene Name: | Rho guanine nucleotide exchange factor 7 |
Gene Aliases: |
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB |
Location: |
Chr.13:111114619-111305734 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF7 | NM_001113511.2 | NP_001106983.1 | ||
| NM_001113512.2 | NP_001106984.1 | |||
| NM_001320852.1 | NP_001307781.1 | |||
| NM_145735.3 | NP_663788.1 | |||
| XM_006719956.3 | XP_006720019.1 | |||
| XM_011521133.2 | XP_011519435.1 | |||
| XM_017020814.1 | XP_016876303.1 | |||
| XM_017020815.1 | XP_016876304.1 | |||
| XM_017020817.1 | XP_016876306.1 | |||
| AB177848.1 | BAD66826.1 | |||
| AL834228.1 | CAD38906.1 | |||
| BC033905.1 | AAH33905.1 | |||
| BC060776.1 | AAH60776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633561 | Chr.13:111116946 - 111122745 on Build GRCh38 | Gain |
|
| dgv373e214 | Chr.13:110983301 - 111125286 on Build GRCh38 | Gain |
|
| nsv1185 | Chr.13:111095621 - 111127624 on Build GRCh38 | Deletion |
|
| esv2759967 | Chr.13:111009686 - 111154564 on Build GRCh38 | Gain |
|
| dgv374e214 | Chr.13:111116844 - 111123840 on Build GRCh38 | Loss |
|
| nsv563182 | Chr.13:111063639 - 111122521 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map