Assay Details
Target Gene Details
Entrez Gene ID: | 79758 |
Gene Name: | dehydrogenase/reductase 12 |
Gene Aliases: |
SDR40C1 |
Location: |
Chr.13:51754856-51804206 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS12 | XM_017020750.1 | XP_016876239.1 | ||
| XM_017020751.1 | XP_016876240.1 |
Target Gene Details
Entrez Gene ID: | 115825 |
Gene Name: | WD repeat and FYVE domain containing 2 |
Gene Aliases: |
PROF, WDF2, ZFYVE22 |
Location: |
Chr.13:51584194-51766799 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY2 | XM_011534914.1 | 12 | 5389 | XP_011533216.1 |
| XM_011534915.2 | 11 | 8197 | XP_011533217.1 | |
| XM_017020386.1 | 12 | 13921 | XP_016875875.1 | |
| AK095358.1 | 1 | 2504 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892343 | Chr.13:51418581 - 52060832 on Build GRCh38 | Gain |
|
| nsv832612 | Chr.13:51602847 - 51788389 on Build GRCh38 | Gain |
|
| dgv3146n54 | Chr.13:51728494 - 51831115 on Build GRCh38 | Loss |
|
| dgv1652n100 | Chr.13:51747428 - 52069788 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map