Assay Details
Target Gene Details
Entrez Gene ID: | 100507240 |
Gene Name: | VWA8 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.13:41961169-41981565 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWA8-AS1 | NR_039974.1 | |||
| AK092199.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3127n54 | Chr.13:41833250 - 42005392 on Build GRCh38 | Loss |
|
| nsv832588 | Chr.13:41876984 - 42053116 on Build GRCh38 | Gain |
|
| dgv75e55 | Chr.13:41819688 - 42008459 on Build GRCh38 | Loss |
|
| nsv1038912 | Chr.13:41930312 - 42030158 on Build GRCh38 | Gain |
|

Genomic Map