Assay Details
Target Gene Details
Entrez Gene ID: | 5101 |
Gene Name: | protocadherin 9 |
Gene Aliases: |
- |
Location: |
Chr.13:66302834-67230336 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH9 | NM_001318372.1 | NP_001305301.1 | ||
| NM_001318373.1 | NP_001305302.1 | |||
| NM_020403.4 | NP_065136.1 | |||
| NM_203487.2 | NP_982354.1 | |||
| XM_017020619.1 | XP_016876108.1 | |||
| AF169692.2 | AAF89689.2 | |||
| AL834258.1 | CAD38933.1 | |||
| BC136626.1 | ||||
| BC136627.1 | ||||
| BC144321.1 | ||||
| BX537422.1 | CAD97664.1 | |||
| KJ534915.1 |
Target Gene Details
Entrez Gene ID: | 100874064 |
Gene Name: | PCDH9 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.13:66825169-66915031 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH9-AS2 | NR_046527.1 | |||
| BX117607.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523360 | Chr.13:66748582 - 66946212 on Build GRCh38 | Loss |
|
| nsv1051576 | Chr.13:66748615 - 66946123 on Build GRCh38 | Loss |
|
| nsv562136 | Chr.13:66781486 - 66911015 on Build GRCh38 | Gain |
|
| nsv1042756 | Chr.13:66778307 - 66871735 on Build GRCh38 | Loss |
|
| nsv562137 | Chr.13:66818166 - 66863184 on Build GRCh38 | Loss |
|
| nsv64575 | Chr.13:66827375 - 66829717 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map