Assay Details
Target Gene Details
Entrez Gene ID: | 100861541 |
Gene Name: | DOCK9 antisense RNA 2 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.13:99086723-99088625 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK9-AS2 | NR_047482.1 | 1 | 617 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892433 | Chr.13:98843569 - 99243711 on Build GRCh38 | Gain |
|
| nsv1050094 | Chr.13:98974062 - 99323969 on Build GRCh38 | Gain |
|
| dgv3361n54 | Chr.13:99086622 - 99087569 on Build GRCh38 | Gain |
|
| nsv1043886 | Chr.13:99038252 - 99112843 on Build GRCh38 | Loss |
|
| nsv562927 | Chr.13:99056285 - 99177057 on Build GRCh38 | Gain |
|
| dgv3362n54 | Chr.13:99086815 - 99087569 on Build GRCh38 | Gain |
|
| esv27384 | Chr.13:99086121 - 99087661 on Build GRCh38 | Loss |
|
| dgv3359n54 | Chr.13:99086152 - 99087408 on Build GRCh38 | Loss |
|

Genomic Map