Assay Details
Target Gene Details
Entrez Gene ID: | 100506394 |
Gene Name: | GAS6 antisense RNA 2 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.13:113864177-113866832 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS6-AS2 | NR_044993.1 | |||
| CA454201.1 | ||||
| DA663147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043348 | Chr.13:113756321 - 113867896 on Build GRCh38 | Gain |
|
| nsv563388 | Chr.13:113754200 - 113872798 on Build GRCh38 | Loss |
|
| nsv832727 | Chr.13:113753115 - 113918257 on Build GRCh38 | Loss |
|
| esv3633626 | Chr.13:113845215 - 113936331 on Build GRCh38 | Gain |
|
| esv3582860 | Chr.13:113756381 - 113867871 on Build GRCh38 | Gain |
|
| nsv563387 | Chr.13:113832338 - 113876058 on Build GRCh38 | Loss |
|
| nsv826836 | Chr.13:113753163 - 113875076 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74118417] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map