Assay Details
Target Gene Details
Entrez Gene ID: | 647174 |
Gene Name: | serpin family E member 3 |
Gene Aliases: |
- |
Location: |
Chr.13:51334976-51362391 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINE3 | XM_017020707.1 | 1 | 5017 | XP_016876196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
| nsv832611 | Chr.13:51192336 - 51364178 on Build GRCh38 | Loss |
|
| nsv1041 | Chr.13:51325852 - 51352515 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map