Assay Details
Target Gene Details
Entrez Gene ID: | 10804 |
Gene Name: | gap junction protein beta 6 |
Gene Aliases: |
CX30, DFNA3, DFNA3B, DFNB1B, ECTD2, ED2, EDH, HED, HED2 |
Location: |
Chr.13:20221962-20232395 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJB6 | NM_001110219.2 | NP_001103689.1 | ||
| NM_001110220.2 | NP_001103690.1 | |||
| NM_001110221.2 | NP_001103691.1 | |||
| XM_017020358.1 | XP_016875847.1 | |||
| XM_017020359.1 | XP_016875848.1 | |||
| XM_017020360.1 | XP_016875849.1 | |||
| AK075247.1 | ||||
| AY789474.1 | AAV67951.1 | |||
| AY789475.1 | AAV67952.1 | |||
| AY789476.1 | AAV67953.1 | |||
| BP227986.1 | ||||
| DA165219.1 | ||||
| DC295753.1 | ||||
| DN998505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631424 | Chr.13:20190147 - 20280383 on Build GRCh38 | Gain |
|
| dgv1612n100 | Chr.13:20222652 - 20530273 on Build GRCh38 | Loss |
|
| dgv3072n54 | Chr.13:20216603 - 20521581 on Build GRCh38 | Loss |
|
| esv3631426 | Chr.13:20219223 - 20261765 on Build GRCh38 | Gain |
|
| nsv1046946 | Chr.13:19730301 - 20507454 on Build GRCh38 | Gain |
|
| esv2751132 | Chr.13:20217943 - 20546665 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map