Assay Details
Target Gene Details
Entrez Gene ID: | 222484 |
Gene Name: | ligand of numb-protein X 2 |
Gene Aliases: |
PDZRN1 |
Location: |
Chr.13:27545913-27621106 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX2 | XM_017020434.1 | XP_016875923.1 | ||
| AK095319.1 |
Target Gene Details
Entrez Gene ID: | 51082 |
Gene Name: | RNA polymerase I subunit D |
Gene Aliases: |
AC19, POLR1C, RPA16, RPA9, RPAC2, RPC16, RPO1-3, TCS2 |
Location: |
Chr.13:27620743-27667422 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR1D | NM_001206559.1 | 1 | 286 | NP_001193488.1 |
| XM_005266412.1 | 1 | 286 | XP_005266469.1 | |
| XM_005266414.2 | 1 | 286 | XP_005266471.1 | |
| XM_017020623.1 | 1 | 242 | XP_016876112.1 | |
| AK307356.1 | 1 | 83 | ||
| DB123903.1 | 1 | 286 | ||
| DR156229.1 | 1 | 258 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039183 | Chr.13:27600204 - 27632386 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map