Assay Details
Target Gene Details
Entrez Gene ID: | 2621 |
Gene Name: | growth arrest specific 6 |
Gene Aliases: |
AXLLG, AXSF |
Location: |
Chr.13:113820549-113864103 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS6 | NM_000820.3 | NP_000811.1 | ||
| AK092028.1 | ||||
| AK122969.1 | ||||
| AK126533.1 | BAC86580.1 | |||
| AK290803.1 | ||||
| AK303145.1 | ||||
| BC038984.1 | AAH38984.1 | |||
| L13720.1 | AAA58494.1 |
Target Gene Details
Entrez Gene ID: | 650669 |
Gene Name: | GAS6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:113815610-113842841 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS6-AS1 | NR_044995.2 | |||
| BX385300.2 | ||||
| DA460725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv563390 | Chr.13:113809317 - 113845944 on Build GRCh38 | Loss |
|
| dgv1135e59 | Chr.13:113824020 - 113824518 on Build GRCh38 | Duplication |
|
| nsv1043348 | Chr.13:113756321 - 113867896 on Build GRCh38 | Gain |
|
| nsv563388 | Chr.13:113754200 - 113872798 on Build GRCh38 | Loss |
|
| nsv951933 | Chr.13:113808028 - 113844427 on Build GRCh38 | Deletion |
|
| nsv832727 | Chr.13:113753115 - 113918257 on Build GRCh38 | Loss |
|
| esv3633615 | Chr.13:113724844 - 113843481 on Build GRCh38 | Gain |
|
| esv24756 | Chr.13:113802553 - 113834906 on Build GRCh38 | Gain+Loss |
|
| esv3582860 | Chr.13:113756381 - 113867871 on Build GRCh38 | Gain |
|
| nsv826836 | Chr.13:113753163 - 113875076 on Build GRCh38 | Loss |
|
| nsv826837 | Chr.13:113802020 - 113834906 on Build GRCh38 | Loss |
|
| esv3633621 | Chr.13:113808937 - 113835264 on Build GRCh38 | Loss |
|
| nsv563391 | Chr.13:113824151 - 113825282 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs67222406] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map