Assay Details
Target Gene Details
Entrez Gene ID: | 160857 |
Gene Name: | coiled-coil domain containing 122 |
Gene Aliases: |
- |
Location: |
Chr.13:43821959-43879690 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC122 | NM_144974.3 | NP_659411.2 | ||
| XM_017020397.1 | XP_016875886.1 | |||
| XM_017020398.1 | XP_016875887.1 | |||
| XM_017020399.1 | XP_016875888.1 | |||
| AK056408.1 | BAB71178.1 | |||
| BC137289.1 | ||||
| BC137290.1 | ||||
| BC144570.1 | ||||
| BC144571.1 | ||||
| DA739121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832595 | Chr.13:43716837 - 43925163 on Build GRCh38 | Gain |
|

Genomic Map