Assay Details
Target Gene Details
Entrez Gene ID: | 9252 |
Gene Name: | ribosomal protein S6 kinase A5 |
Gene Aliases: |
MSK1, MSPK1, RLPK |
Location: |
Chr.14:90868122-91060649 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KA5 | NM_001322227.1 | 18 | 6514 | NP_001309156.1 |
| NM_001322228.1 | 16 | 6246 | NP_001309157.1 | |
| NM_001322229.1 | 17 | 6333 | NP_001309158.1 | |
| NM_001322230.1 | 18 | 6541 | NP_001309159.1 | |
| NM_001322231.1 | 16 | 6250 | NP_001309160.1 | |
| NM_001322232.1 | 16 | 6135 | NP_001309161.1 | |
| NM_001322233.1 | 16 | 6192 | NP_001309162.1 | |
| NM_001322234.1 | 17 | 6391 | NP_001309163.1 | |
| NM_001322235.1 | 18 | 6494 | NP_001309164.1 | |
| NM_001322236.1 | 16 | 6270 | NP_001309165.1 | |
| NM_001322237.1 | 17 | 6153 | NP_001309166.1 | |
| NM_001322238.1 | 15 | 5862 | NP_001309167.1 | |
| NM_004755.3 | 17 | 6354 | NP_004746.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832855 | Chr.14:90723955 - 90927660 on Build GRCh38 | Loss |
|
| nsv832856 | Chr.14:90846980 - 91031317 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map