Assay Details
Target Gene Details
Entrez Gene ID: | 319089 |
Gene Name: | tetratricopeptide repeat domain 6 |
Gene Aliases: |
C14orf25, NCRNA00291 |
Location: |
Chr.14:37595199-37842628 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC6 | NM_001310135.1 | NP_001297064.1 | ||
| XM_011537432.2 | XP_011535734.1 | |||
| XM_017021254.1 | XP_016876743.1 | |||
| XM_017021255.1 | XP_016876744.1 | |||
| XM_017021257.1 | XP_016876746.1 | |||
| BC038110.2 | AAH38110.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832772 | Chr.14:37561095 - 37756716 on Build GRCh38 | Loss |
|
| nsv1054995 | Chr.14:37608822 - 37668971 on Build GRCh38 | Loss |
|
| nsv826923 | Chr.14:37548414 - 37639440 on Build GRCh38 | Gain |
|

Genomic Map