Assay Details
Target Gene Details
Entrez Gene ID: | 8846 |
Gene Name: | alkB homolog 1, histone H2A dioxygenase |
Gene Aliases: |
ABH, ABH1, ALKBH, alkB, hABH |
Location: |
Chr.14:77672404-77708038 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALKBH1 | NM_006020.2 | NP_006011.2 | ||
| BC015024.1 | AAH15024.1 | |||
| BC025787.1 | AAH25787.1 | |||
| X91992.1 | CAA63047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635010 | Chr.14:77630281 - 77745656 on Build GRCh38 | Gain |
|
| esv34940 | Chr.14:77638554 - 77756455 on Build GRCh38 | Gain |
|
| nsv1053810 | Chr.14:77521528 - 77681977 on Build GRCh38 | Gain |
|
| nsv521287 | Chr.14:77641660 - 77744882 on Build GRCh38 | Gain |
|
| esv3635012 | Chr.14:77644082 - 77697186 on Build GRCh38 | Gain |
|
| esv2759998 | Chr.14:77607837 - 77796229 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72685244] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map