Assay Details
Target Gene Details
Entrez Gene ID: | 647286 |
Gene Name: | retinal degeneration 3-like |
Gene Aliases: |
TDRD9-AS1, TDRD9AS1 |
Location: |
Chr.14:103940426-103942308 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RD3L | NM_001257268.1 | NP_001244197.1 | ||
| BQ184214.1 | ||||
| BQ185906.1 | ||||
| BX117646.1 |
Target Gene Details
Entrez Gene ID: | 122402 |
Gene Name: | tudor domain containing 9 |
Gene Aliases: |
C14orf75, HIG-1, NET54 |
Location: |
Chr.14:103928343-104052667 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDRD9 | NM_153046.2 | NP_694591.2 | ||
| XM_005267309.3 | XP_005267366.1 | |||
| XM_006720019.3 | XP_006720082.1 | |||
| XM_006720020.3 | XP_006720083.1 | |||
| XM_011536397.2 | XP_011534699.1 | |||
| XM_011536398.2 | XP_011534700.1 | |||
| XM_011536400.2 | XP_011534702.1 | |||
| XM_017020955.1 | XP_016876444.1 | |||
| BG719020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
| esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map