Assay Details
Target Gene Details
Entrez Gene ID: | 91828 |
Gene Name: | exocyst complex component 3 like 4 |
Gene Aliases: |
C14orf73 |
Location: |
Chr.14:103094202-103110562 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOC3L4 | NM_001077594.1 | NP_001071062.1 | ||
| XM_011537323.2 | XP_011535625.1 | |||
| XM_011537324.2 | XP_011535626.1 | |||
| XM_011537325.2 | XP_011535627.1 | |||
| XM_011537327.2 | XP_011535629.1 | |||
| XM_011537328.2 | XP_011535630.1 | |||
| XM_011537329.2 | XP_011535631.1 | |||
| XM_011537330.2 | XP_011535632.1 | |||
| XM_011537331.1 | XP_011535633.1 | |||
| XM_011537332.2 | XP_011535634.1 | |||
| XM_011537333.2 | XP_011535635.1 | |||
| XM_011537334.1 | XP_011535636.1 | |||
| BC113655.1 | ||||
| BC117375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565970 | Chr.14:103089160 - 103139470 on Build GRCh38 | Loss |
|
| nsv565971 | Chr.14:103100567 - 103135315 on Build GRCh38 | Loss |
|
| nsv9168 | Chr.14:103065392 - 103446969 on Build GRCh38 | Loss |
|
| nsv832882 | Chr.14:102970609 - 103142844 on Build GRCh38 | Loss |
|
| nsv952257 | Chr.14:103067564 - 103135963 on Build GRCh38 | Deletion |
|
| esv3635574 | Chr.14:102931859 - 103104526 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map