Assay Details
Target Gene Details
Entrez Gene ID: | 1965 |
Gene Name: | eukaryotic translation initiation factor 2 subunit alpha |
Gene Aliases: |
EIF-2, EIF-2A, EIF-2alpha, EIF2, EIF2A |
Location: |
Chr.14:67360317-67386516 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2S1 | NM_004094.4 | NP_004085.1 | ||
| AK297244.1 | ||||
| AK312442.1 | ||||
| BC002513.2 | AAH02513.1 | |||
| BX161521.1 | CAD61953.1 | |||
| J02645.1 | AAA52373.1 |
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPHN | XM_017020925.1 | XP_016876414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751285 | Chr.14:66898030 - 67420064 on Build GRCh38 | Gain |
|
| nsv564961 | Chr.14:67266144 - 67637063 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map