Assay Details
Target Gene Details
Entrez Gene ID: | 51077 |
Gene Name: | FCF1 rRNA-processing protein |
Gene Aliases: |
Bka, C14orf111, CGI-35, UTP24 |
Location: |
Chr.14:74713118-74738620 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCF1 | NM_001318508.1 | NP_001305437.1 | ||
| NM_015962.4 | NP_057046.1 | |||
| XM_005267730.3 | XP_005267787.1 | |||
| XM_011536815.2 | XP_011535117.1 | |||
| XM_011536816.2 | XP_011535118.1 | |||
| AF132969.1 | AAD27744.1 | |||
| AK290666.1 | ||||
| BC080600.1 | AAH80600.1 | |||
| BX248271.1 | CAD62599.1 | |||
| BX647810.1 | CAJ18322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832832 | Chr.14:74706738 - 74884573 on Build GRCh38 | Gain+Loss |
|
| nsv832831 | Chr.14:74546343 - 74740664 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map