Assay Details
Target Gene Details
Entrez Gene ID: | 253970 |
Gene Name: | surfactant associated 3 |
Gene Aliases: |
NANCI, SFTPH, SP-H |
Location: |
Chr.14:36473288-36513785 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFTA3 | NM_001101341.1 | NP_001094811.1 | ||
| AK027147.1 | 1 | 221 | ||
| AY102071.1 | ||||
| KF856952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832771 | Chr.14:36301801 - 36494836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map