Assay Details
Target Gene Details
Entrez Gene ID: | 100129794 |
Gene Name: | SLC25A21 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.14:37172026-37173794 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A21-AS1 | NR_033240.1 | 1 | 1402 | |
| AK057476.1 | 1 | 1325 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1871n100 | Chr.14:37087152 - 37201854 on Build GRCh38 | Gain |
|
| nsv1153559 | Chr.14:37162398 - 37302029 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117763002] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map