Assay Details
Target Gene Details
Entrez Gene ID: | 283554 |
Gene Name: | G protein-coupled receptor 137C |
Gene Aliases: |
TM7SF1L2 |
Location: |
Chr.14:52553148-52637713 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR137C | NM_001099652.1 | NP_001093122.1 | ||
| AK123953.1 | ||||
| AL834372.1 | CAD39035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826955 | Chr.14:52561923 - 52565098 on Build GRCh38 | Loss |
|
| esv2670271 | Chr.14:52559510 - 52564934 on Build GRCh38 | Deletion |
|
| esv2748718 | Chr.14:52559523 - 52564928 on Build GRCh38 | Deletion |
|
| esv3634524 | Chr.14:52559500 - 52564925 on Build GRCh38 | Loss |
|

Genomic Map