Assay Details
Target Gene Details
Entrez Gene ID: | 90381 |
Gene Name: | TOPBP1 interacting checkpoint and replication regulator |
Gene Aliases: |
C15orf42, SLD3, Treslin |
Location: |
Chr.15:89575587-89628023 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TICRR | NM_001308025.1 | NP_001294954.1 | ||
| NM_152259.3 | NP_689472.3 | |||
| CN285632.1 | ||||
| GQ227787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892721 | Chr.15:89530925 - 89673865 on Build GRCh38 | Gain |
|
| nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain |
|
| esv34025 | Chr.15:89412131 - 89630626 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map