Assay Details
Target Gene Details
Entrez Gene ID: | 6263 |
Gene Name: | ryanodine receptor 3 |
Gene Aliases: |
RYR-3 |
Location: |
Chr.15:33310773-33866103 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RYR3 | NM_001036.4 | NP_001027.3 | ||
| NM_001243996.2 | NP_001230925.1 | |||
| XM_011521880.2 | XP_011520182.1 | |||
| XM_017022468.1 | XP_016877957.1 | |||
| XM_017022469.1 | XP_016877958.1 | |||
| XM_017022471.1 | XP_016877960.1 | |||
| XM_017022472.1 | XP_016877961.1 | |||
| XM_017022473.1 | XP_016877962.1 | |||
| XM_017022475.1 | XP_016877964.1 | |||
| XM_017022476.1 | XP_016877965.1 | |||
| XM_017022477.1 | XP_016877966.1 | |||
| AB001025.1 | BAA23795.1 | |||
| AJ001515.1 | CAA04798.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832964 | Chr.15:33316570 - 33491817 on Build GRCh38 | Loss |
|
| nsv568919 | Chr.15:33346134 - 33444482 on Build GRCh38 | Loss |
|
| nsv1052620 | Chr.15:32612402 - 34538731 on Build GRCh38 | Gain |
|
| nsv832963 | Chr.15:33222597 - 33420241 on Build GRCh38 | Gain |
|
| esv33337 | Chr.15:31689863 - 34575097 on Build GRCh38 | Gain+Loss |
|
| dgv4456n54 | Chr.15:32604149 - 34557061 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74368065] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map