Assay Details
Target Gene Details
Entrez Gene ID: | 8120 |
Gene Name: | adaptor related protein complex 3 beta 2 subunit |
Gene Aliases: |
NAPTB |
Location: |
Chr.15:82659281-82710112 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3B2 | NM_001278511.1 | NP_001265440.1 | ||
| NM_001278512.1 | NP_001265441.1 | |||
| NM_004644.4 | NP_004635.2 | |||
| XM_017022640.1 | XP_016878129.1 | |||
| XM_017022641.1 | XP_016878130.1 | |||
| AF022152.1 | AAB71894.1 | |||
| AK097827.1 | ||||
| BC093739.1 | AAH93739.1 | |||
| BC143342.1 | ||||
| BC143346.1 | ||||
| CR749860.1 | CAH18704.2 | |||
| U37673.1 | AAC50219.1 |
Target Gene Details
Entrez Gene ID: | 283692 |
Gene Name: | CPEB1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:82647770-82692820 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB1-AS1 | NR_046096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2637n100 | Chr.15:82568764 - 82803755 on Build GRCh38 | Gain |
|
| esv3637058 | Chr.15:82590287 - 82726611 on Build GRCh38 | Gain |
|
| nsv817702 | Chr.15:82630710 - 82788672 on Build GRCh38 | Loss |
|
| esv34318 | Chr.15:82614804 - 82784743 on Build GRCh38 | Loss |
|
| esv2674021 | Chr.15:82628241 - 82789040 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map