Assay Details
Target Gene Details
Entrez Gene ID: | 64506 |
Gene Name: | cytoplasmic polyadenylation element binding protein 1 |
Gene Aliases: |
CPE-BP1, CPEB, CPEB-1, h-CPEB, hCPEB-1 |
Location: |
Chr.15:82543201-82648016 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB1 | NM_030594.4 | 1 | 72 | NP_085097.3 |
| BC050629.1 | AAH50629.1 | |||
| HY007452.1 | 1 | 73 |
Target Gene Details
Entrez Gene ID: | 283692 |
Gene Name: | CPEB1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:82647770-82692820 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB1-AS1 | NR_046096.1 | 1 | 176 | |
| DB070426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2637n100 | Chr.15:82568764 - 82803755 on Build GRCh38 | Gain |
|
| esv3637058 | Chr.15:82590287 - 82726611 on Build GRCh38 | Gain |
|
| nsv817702 | Chr.15:82630710 - 82788672 on Build GRCh38 | Loss |
|
| esv33057 | Chr.15:82646082 - 82648837 on Build GRCh38 | Gain |
|
| esv34318 | Chr.15:82614804 - 82784743 on Build GRCh38 | Loss |
|
| esv2674021 | Chr.15:82628241 - 82789040 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map