Assay Details
Target Gene Details
Entrez Gene ID: | 2923 |
Gene Name: | protein disulfide isomerase family A member 3 |
Gene Aliases: |
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC |
Location: |
Chr.15:43746392-43772606 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDIA3 | NM_005313.4 | NP_005304.3 | ||
AK075455.1 | ||||
AK075481.1 | ||||
AK293249.1 | ||||
BC014433.1 | AAH14433.1 | |||
BC036000.1 | ||||
BC071878.1 | AAH71878.1 | |||
D16234.1 | BAA03759.1 | |||
D83485.1 | BAA11928.1 | |||
EU668338.1 | ||||
FJ224330.1 | ||||
U42068.1 | AAC50331.1 | |||
Z49835.1 | CAA89996.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1036318 | Chr.15:43761419 - 43868575 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2760028 | Chr.15:43530960 - 43856443 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv33661 | Chr.15:43724321 - 43772222 on Build GRCh38 | Loss |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
