Assay Details
Target Gene Details
Entrez Gene ID: | 6557 |
Gene Name: | solute carrier family 12 member 1 |
Gene Aliases: |
BSC1, NKCC2 |
Location: |
Chr.15:48206301-48304078 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC12A1 | NM_000338.2 | NP_000329.2 | ||
| NM_001184832.1 | NP_001171761.1 | |||
| XM_005254606.1 | XP_005254663.1 | |||
| AK295375.1 | ||||
| AK298275.1 | ||||
| AK298312.1 | ||||
| BC040138.2 | AAH40138.1 | |||
| BX647067.1 | ||||
| BX647484.1 | ||||
| EF559316.2 | ||||
| U58130.1 | AAB07364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522923 | Chr.15:48228393 - 48237743 on Build GRCh38 | Loss |
|
| esv2760030 | Chr.15:48216841 - 48371581 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map